Assay Details
| Gene Symbol: Supt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019995.3 | NP_064379.1 | 3 - 4 | 226 | 70 | |
| GenBank mRNA | AF139179.1 | - | 3 - 4 | 230 | 70 | |
| AK050619.1 | - | 4 - 5 | 253 | 70 | ||
| AK083379.1 | - | 4 - 5 | 253 | 70 | ||
| AK212629.1 | - | 2 - 3 | 83 | 70 | ||
| BC052702.1 | - | 3 - 4 | 218 | 6400922 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF139179.1, AK050619.1, AK083379.1, BC052702.1, NM_019995.3 transcripts.
Set Membership: |
|

Supt20
Genomic Map