Assay Details
| Gene Symbol: Supt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK171787.1 | - | 6 - 7 | 1458 | 58 | |
| BC052702.1 | - | 23 - 24 | 3097 | 6400922 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Supt20
Genomic Map