Assay Details
| Gene Symbol: Pde7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122759.2 | NP_001116231.1 | 3 - 4 | 599 | 68 | |
| NM_008802.3 | NP_032828.2 | 3 - 4 | 329 | 68 | ||
| GenBank mRNA | AK154521.1 | - | 3 - 4 | 416 | 68 | |
| AK168957.1 | - | 3 - 4 | 291 | 68 | ||
| AY007702.1 | - | 3 - 4 | 278 | 68 | ||
| BC062909.1 | - | 3 - 4 | 598 | 6815827 | 68 | |
| CV558025.1 | - | 3 - 4 | 329 | 68 | ||
| U68171.1 | - | 3 - 4 | 277 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde7a
Genomic Map