Assay Details
| Gene Symbol: Arhgef26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081295.1 | NP_001074764.1 | 11 - 12 | 3014 | 71 | |
| GenBank mRNA | AK144976.1 | - | 11 - 12 | 2520 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef26
Genomic Map