Assay Details
| Gene Symbol: Arhgef26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081295.1 | NP_001074764.1 | 2 - 3 | 1843 | 116 | |
| GenBank mRNA | AK087060.1 | - | 2 - 3 | 1623 | 117 | |
| AK144976.1 | - | 2 - 3 | 1349 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef26
Genomic Map