Assay Details
| Gene Symbol: Arhgef26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081295.1 | NP_001074764.1 | 3 - 4 | 1984 | 107 | |
| GenBank mRNA | AK087060.1 | - | 3 - 4 | 1766 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef26
Genomic Map