Assay Details
Gene Symbol: Zfp639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144519.4 | NP_653102.1 | 1 - 2 | 141 | 112 | |
GenBank mRNA | AK076145.1 | - | 1 - 2 | 142 | 112 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK076145.1, NM_144519.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
