Assay Details
| Gene Symbol: Zfp639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161818.1 | NP_001155290.1 | 2 - 3 | 226 | 70 | |
| NM_144519.4 | NP_653102.1 | 2 - 3 | 327 | 70 | ||
| GenBank mRNA | AK007493.1 | - | 2 - 3 | 204 | 70 | |
| AK049934.1 | - | 2 - 3 | 206 | 70 | ||
| AK076145.1 | - | 2 - 3 | 328 | 70 | ||
| AK078000.1 | - | 2 - 3 | 227 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007493.1, AK049934.1, AK076145.1, AK078000.1, NM_001161818.1, NM_144519.4 transcripts.
Set Membership: |
|

Zfp639
Genomic Map