Assay Details
Gene Symbol: Zfp639 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161818.1 | NP_001155290.1 | 4 - 5 | 368 | 150 | |
NM_144519.4 | NP_653102.1 | 4 - 5 | 469 | 150 | ||
GenBank mRNA | AK049934.1 | - | 4 - 5 | 348 | 150 | |
AK076145.1 | - | 4 - 5 | 470 | 150 | ||
AK076837.1 | - | 4 - 5 | 345 | 150 | ||
AK078000.1 | - | 4 - 5 | 369 | 150 | ||
BC003941.1 | - | 2 - 3 | 118 | 3487151 | 150 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK049934.1, AK076145.1, AK076837.1, AK078000.1, BC003941.1, NM_001161818.1, NM_144519.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
