Assay Details
| Gene Symbol: Slc33a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272035.1 | NP_001258964.1 | 2 - 3 | 1515 | 65 | |
| NM_015728.5 | NP_056543.2 | 2 - 3 | 1515 | 65 | ||
| GenBank mRNA | AB016795.1 | - | 2 - 3 | 972 | 65 | |
| AK144703.1 | - | 2 - 3 | 1382 | 65 | ||
| BC005616.1 | - | 2 - 3 | 1092 | 3154886 | 65 | |
| BC054395.1 | - | 2 - 3 | 1515 | 6492236 | 65 | |
| BC063104.1 | - | 1448 | 5042025 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc33a1
Genomic Map