Assay Details
| Gene Symbol: Eif2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005509.2 | NP_001005509.1 | 11 - 12 | 1736 | 107 | |
| GenBank mRNA | AK050971.1 | - | 11 - 12 | 1453 | 107 | |
| AK078519.1 | - | 11 - 12 | 1515 | 107 | ||
| BC030447.1 | - | 10 - 11 | 1405 | 5400637 | 107 | |
| BC082557.1 | - | 11 - 12 | 1541 | 30606936 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif2a
Genomic Map