Assay Details
| Gene Symbol: Eif2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005509.2 | NP_001005509.1 | 7 - 8 | 790 | 106 | |
| GenBank mRNA | AK044865.1 | - | 1 - 2 | 837 | 106 | |
| AK050971.1 | - | 7 - 8 | 507 | 106 | ||
| AK078519.1 | - | 7 - 8 | 569 | 106 | ||
| AK135515.1 | - | 1 - 2 | 1788 | 106 | ||
| AK145275.1 | - | 7 - 8 | 746 | 106 | ||
| AK161904.1 | - | 7 - 8 | 571 | 106 | ||
| BC030447.1 | - | 6 - 7 | 459 | 5400637 | 106 | |
| BC082557.1 | - | 7 - 8 | 595 | 30606936 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif2a
Genomic Map