Assay Details
| Gene Symbol: Atp5s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026536.1 | NP_080812.1 | 4 - 5 | 545 | 148 | |
| GenBank mRNA | AK003725.1 | - | 4 - 5 | 545 | 148 | |
| AK009141.1 | - | 5 - 6 | 657 | 148 | ||
| BC027442.1 | - | 4 - 5 | 527 | 4015414 | 148 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28516081 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28516083 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28516082 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195798 | NM_026536.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195798 | AK003725.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195798 | AK009141.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195798 | BC027442.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195798 | NM_026536.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195798 | AK003725.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195798 | AK009141.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P195798 | BC027442.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 294176 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904398 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 427184 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 742591 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 484052 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 672598 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 563855 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 347386 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333231 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 716505 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 153030 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 126593 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168742 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159892 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195798 | NM_026536.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195798 | AK003725.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195798 | AK009141.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195798 | BC027442.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195798 | NM_026536.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195798 | AK003725.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195798 | AK009141.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P195798 | BC027442.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P195798 | NM_026536.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P195798 | AK003725.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P195798 | AK009141.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P195798 | BC027442.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P195798 | NM_026536.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P195798 | AK003725.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P195798 | AK009141.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P195798 | BC027442.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P192654 | NM_026536.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P192654 | AK003725.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28516092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28516081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28516083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28516086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28516095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28516082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28516094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28516087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28516093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28516084 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07655122 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07655145 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168742 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159892 | - | - |
| Whole Mouse Genome G4122A | A_51_P192654 | NM_026536.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192654 | AK003725.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195798 | NM_026536.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195798 | AK003725.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195798 | AK009141.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195798 | BC027442.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195798 | NM_026536.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195798 | AK003725.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195798 | AK009141.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P195798 | BC027442.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113415 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139764 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159892 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168742 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10122132 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28516083 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28516082 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28516081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28516099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10402205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516113 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10006871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09861436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10289735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516084 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28516136 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10396114 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10396115 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10396116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10396117 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10396113 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417970_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10396112 | - | - |
| Mouse Genome 430 2.0 Array | 1417970_at | - | - |
| Mouse Genome 430 2.0 Array | 1417970_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68055_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68055_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200003854.mm.1 | NM_026536.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003854.mm.1 | NM_026536.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003854.mm.1 | TC1200000576.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417970_PM_at | - | - |
| Mouse Expression Set 430 | 1417970_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417970_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417970_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417970_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417970_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 416391 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4200139 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104780746 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4200139 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004200139 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2642158 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2642158 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230199 | - | - |
| mouse-6 v1.1 (Illumina) | scl0002312.1_1-S | - | - |
| mouse-6 v1.1 (Illumina) | scl43153.6_180-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104780746 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4200139 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl43153.6_180-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104780746 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4200139 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2642158 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1230199 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2642158 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl43153.6_180-S | - | - |

Atp5s
Genomic Map