Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200013002.mm.1 |
NM_013896.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013002.mm.1 |
NM_013896.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200013002.mm.1 |
TC1200001893.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200012999.mm.1 |
NM_001024853.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200012999.mm.1 |
NM_001024853.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200012999.mm.1 |
NM_001024854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200012999.mm.1 |
NM_001024854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200012999.mm.1 |
NM_001286203.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200012999.mm.1 |
NM_013896.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200012999.mm.1 |
NM_013896.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200012999.mm.1 |
TC1200001893.mm.1 |
MTA TC |