Assay Details
| Gene Symbol: Timm9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024853.1 | NP_001020024.1 | 1 - 2 | 120 | 120 | |
| GenBank mRNA | BY100887.1 | - | 1 - 2 | 119 | 120 | |
| DV652207.1 | - | 1 - 2 | 97 | 120 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001024853.1 transcripts.
Set Membership: |
|

Timm9
Genomic Map