Assay Details
| Gene Symbol: Shroom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027917.3 | NP_082193.2 | 3 - 4 | 1160 | 84 | |
| GenBank mRNA | AK004934.1 | - | 3 - 4 | 1087 | 84 | |
| AK134332.1 | - | 3 - 4 | 1088 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Shroom1
Genomic Map