Assay Details
| Gene Symbol: Mrps36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190264.1 | NP_001177193.1 | 2 - 3 | 190 | 128 | |
| GenBank mRNA | AK009966.1 | - | 2 - 3 | 121 | 128 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29269915 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29269917 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29269918 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29269919 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29269913 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29269914 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166528 | AK003111.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166528 | AK003765.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166528 | NM_001190264.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166528 | NM_025369.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2125561 | AK007512.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2125561 | AK009050.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2125561 | AK009966.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2125561 | AK003111.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2125561 | AK003765.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 178979 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 7643 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 431939 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355777 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 824000 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178026 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2125561 | AK007512.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2125561 | AK009050.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2125561 | AK009966.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2125561 | AK003111.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2125561 | AK003765.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166528 | AK003111.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166528 | AK003765.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166528 | NM_001190264.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166528 | NM_025369.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29269927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29269912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29269925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29269913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29269922 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29269921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29269926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29269932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29269933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29269904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29269906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29269930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29269909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29269911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29269919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29269910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29269917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29269907 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178026 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07973012 | - | - |
| Whole Mouse Genome G4122A | A_51_P190697 | AK007512.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190697 | AK009050.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190697 | AK009966.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190697 | AK003111.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190697 | AK003765.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190690 | AK003111.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190690 | AK003765.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190690 | NM_001190264.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190690 | NM_025369.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136171 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178026 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10451902 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10451912 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29269918 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29269915 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29269917 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29269913 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29269914 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29269919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29269945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29269934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29269935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29269919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29269937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29269941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10617523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29269928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29269930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29269939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29269932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29269931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29269925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29269922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10534626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29269938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29269933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29269921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11099997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10710488 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29269936 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10411724 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411727 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411725 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411723 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566515 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411726 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423242_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10566514 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10411722 | - | - |
| Mouse Genome 430 2.0 Array | 1423242_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423242_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300017316.mm.1 | NM_001190264.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300017316.mm.1 | NM_001190264.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300017316.mm.1 | NM_025369.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300017316.mm.1 | NM_025369.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300017316.mm.1 | TC1300002541.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300017315.mm.1 | NM_001190264.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300017315.mm.1 | NM_001190264.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300017315.mm.1 | NM_025369.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300017315.mm.1 | NM_025369.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300017315.mm.1 | TC1300002541.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300017308.mm.1 | NM_001190264.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300017308.mm.1 | NM_001190264.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300017308.mm.1 | NM_025369.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300017308.mm.1 | NM_025369.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300017308.mm.1 | TC1300002541.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300017317.mm.1 | NM_001190264.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300017317.mm.1 | NM_001190264.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300017317.mm.1 | NM_025369.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300017317.mm.1 | NM_025369.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300017317.mm.1 | TC1300002541.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423242_PM_at | - | - |
| Mouse Expression Set 430 | 1423242_at | - | - |
| Murine Genome U74v2 Set | 98929_at | - | - |
| Murine Genome U74v2 Set | 98929_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423242_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423242_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423242_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423242_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 659915 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 7050072 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 7050072 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007050072 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2681528 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2681528 | - | - |
| mouse-6 v1.1 (Illumina) | scl0066128.1_169-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7050072 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0066128.1_169-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7050072 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2681528 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2681528 | - | - |

Mrps36
Genomic Map