Assay Details
| Gene Symbol: Ankra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271390.1 | NP_001258319.1 | 1 - 2 | 404 | 87 | |
| NM_001271391.1 | NP_001258320.1 | 1 - 2 | 404 | 87 | ||
| NM_001271392.1 | NP_001258321.1 | 1 - 2 | 404 | 87 | ||
| GenBank mRNA | AK003438.2 | - | 1 - 2 | 323 | 87 | |
| AK081695.1 | - | 1 - 2 | 289 | 87 | ||
| BC037437.1 | - | 1 - 2 | 332 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK081695.1, NM_001271390.1, NM_001271391.1, NM_001271392.1 transcripts.
Set Membership: |
|

Ankra2
Genomic Map