Assay Details
| Gene Symbol: Msh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010829.2 | NP_034959.2 | 10 - 11 | 1545 | 112 | |
| GenBank mRNA | AK083388.1 | - | 11 - 12 | 1858 | 112 | |
| AK133907.1 | - | 10 - 11 | 1537 | 112 | ||
| AK161089.1 | - | 10 - 11 | 1541 | 112 | ||
| BC150759.1 | - | 10 - 11 | 1464 | 9087670 | 112 | |
| BQ571929.1 | - | 4 - 5 | 551 | 112 | ||
| M80360.1 | - | 10 - 11 | 1542 | 112 | ||
More Information
Set Membership: |
|

Msh3
Genomic Map