Assay Details
| Gene Symbol: Epb4.1l5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113416.1 | NP_001106887.1 | 3 - 4 | 503 | 159 | |
| NM_145506.4 | NP_663481.2 | 3 - 4 | 503 | 159 | ||
| GenBank mRNA | AK034676.1 | - | 3 - 4 | 425 | 159 | |
| AK040354.1 | - | 3 - 4 | 471 | 159 | ||
| AK044959.1 | - | 3 - 4 | 441 | 159 | ||
| AK044996.1 | - | 3 - 4 | 449 | 159 | ||
| AK047523.1 | - | 3 - 4 | 430 | 159 | ||
| AK079301.1 | - | 3 - 4 | 454 | 159 | ||
| AK136125.1 | - | 3 - 4 | 458 | 159 | ||
| AK161141.1 | - | 3 - 4 | 454 | 159 | ||
| AK173198.1 | - | 3 - 4 | 437 | 159 | ||
| BC003937.1 | - | 3 - 4 | 488 | 3486543 | 159 | |
| BC011476.1 | - | 3 - 4 | 437 | 4014009 | 159 | |
More Information
Updated Mapping Information
Set Membership: |
|

Epb4.1l5
Genomic Map