Assay Details
| Gene Symbol: Mcm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008567.1 | NP_032593.1 | 2 - 3 | 346 | 74 | |
| GenBank mRNA | AK088493.1 | - | 2 - 3 | 345 | 74 | |
| AK145520.1 | - | 2 - 3 | 337 | 74 | ||
| AK146272.1 | - | 2 - 3 | 343 | 74 | ||
| AK146770.1 | - | 2 - 3 | 338 | 74 | ||
| AK146896.1 | - | 2 - 3 | 321 | 74 | ||
| AK150726.1 | - | 2 - 3 | 294 | 74 | ||
| AK177954.1 | - | 2 - 3 | 181 | 74 | ||
| AK180079.1 | - | 2 - 3 | 209 | 74 | ||
| AK188642.1 | - | 1 - 2 | 169 | 74 | ||
| AK194535.1 | - | 2 - 3 | 209 | 74 | ||
| AK210308.1 | - | 1 - 2 | 161 | 74 | ||
| AK210474.1 | - | 2 - 3 | 208 | 74 | ||
| AK211618.1 | - | 2 - 3 | 208 | 74 | ||
| BC050886.1 | - | 2 - 3 | 354 | 5703922 | 74 | |
| BC051658.1 | - | 2 - 3 | 323 | 74 | ||
| BC057584.1 | - | 2 - 3 | 324 | 6401827 | 74 | |
| D86726.1 | - | 2 - 3 | 346 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcm6
Genomic Map