Assay Details
| Gene Symbol: Mcm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008567.1 | NP_032593.1 | 3 - 4 | 448 | 124 | |
| GenBank mRNA | AK088493.1 | - | 3 - 4 | 447 | 124 | |
| AK145520.1 | - | 3 - 4 | 439 | 124 | ||
| AK146272.1 | - | 3 - 4 | 445 | 124 | ||
| AK146770.1 | - | 3 - 4 | 440 | 124 | ||
| AK146896.1 | - | 3 - 4 | 423 | 124 | ||
| AK150726.1 | - | 3 - 4 | 396 | 124 | ||
| AK180079.1 | - | 3 - 4 | 311 | 124 | ||
| AK188642.1 | - | 2 - 3 | 271 | 124 | ||
| AK210308.1 | - | 2 - 3 | 124 | |||
| AK210474.1 | - | 3 - 4 | 310 | 124 | ||
| AK211618.1 | - | 3 - 4 | 310 | 124 | ||
| BC050886.1 | - | 3 - 4 | 456 | 5703922 | 124 | |
| BC051658.1 | - | 3 - 4 | 425 | 124 | ||
| BC057584.1 | - | 3 - 4 | 426 | 6401827 | 124 | |
| D86726.1 | - | 3 - 4 | 448 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcm6
Genomic Map