Assay Details
| Gene Symbol: Mcm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008567.1 | NP_032593.1 | 8 - 9 | 1313 | 136 | |
| GenBank mRNA | AK078126.1 | - | 2 - 3 | 307 | 136 | |
| AK088493.1 | - | 8 - 9 | 1312 | 136 | ||
| AK145520.1 | - | 8 - 9 | 1304 | 136 | ||
| AK146272.1 | - | 8 - 9 | 1310 | 136 | ||
| AK146770.1 | - | 8 - 9 | 1306 | 136 | ||
| AK146896.1 | - | 8 - 9 | 1188 | 136 | ||
| AK150726.1 | - | 8 - 9 | 1261 | 136 | ||
| BC050886.1 | - | 8 - 9 | 1321 | 5703922 | 136 | |
| BC051658.1 | - | 8 - 9 | 1288 | 134 | ||
| BC057584.1 | - | 8 - 9 | 1291 | 6401827 | 136 | |
| D86726.1 | - | 8 - 9 | 1313 | 136 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcm6
Genomic Map