Assay Details
| Gene Symbol: Mcm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008567.1 | NP_032593.1 | 9 - 10 | 1458 | 71 | |
| GenBank mRNA | AK078126.1 | - | 3 - 4 | 452 | 71 | |
| AK088493.1 | - | 9 - 10 | 1457 | 71 | ||
| AK145520.1 | - | 9 - 10 | 1449 | 71 | ||
| AK146272.1 | - | 9 - 10 | 1455 | 71 | ||
| AK146770.1 | - | 9 - 10 | 1451 | 71 | ||
| AK146896.1 | - | 9 - 10 | 1333 | 71 | ||
| AK150726.1 | - | 9 - 10 | 1406 | 71 | ||
| BC050886.1 | - | 9 - 10 | 1466 | 5703922 | 71 | |
| BC051658.1 | - | 9 - 10 | 1433 | 71 | ||
| BC057584.1 | - | 9 - 10 | 1436 | 6401827 | 71 | |
| D86726.1 | - | 9 - 10 | 1458 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mcm6
Genomic Map