Assay Details
| Gene Symbol: Serpinb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009257.3 | NP_033283.1 | 3 - 4 | 382 | 127 | |
| GenBank mRNA | AK136532.1 | - | 3 - 4 | 388 | 127 | |
| AK145220.1 | - | 2 - 3 | 350 | 127 | ||
| U54705.1 | - | 4 - 5 | 489 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Serpinb5
Genomic Map