Assay Details
| Gene Symbol: Serpinb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009257.3 | NP_033283.1 | 4 - 5 | 508 | 114 | |
| GenBank mRNA | AK136532.1 | - | 4 - 5 | 514 | 114 | |
| AK145220.1 | - | 3 - 4 | 476 | 114 | ||
| BC005434.1 | - | 4 - 5 | 509 | 3600282 | 114 | |
| U54705.1 | - | 5 - 6 | 615 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Serpinb5
Genomic Map