Assay Details
| Gene Symbol: Prkch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008856.3 | NP_032882.2 | 3 - 4 | 698 | 66 | |
| GenBank mRNA | AK076352.1 | - | 3 - 4 | 897 | 66 | |
| AK145817.1 | - | 3 - 4 | 944 | 66 | ||
| AK155951.1 | - | 4 - 5 | 1041 | 66 | ||
| AK164044.1 | - | 3 - 4 | 897 | 66 | ||
| BC031121.1 | - | 3 - 4 | 698 | 5059633 | 66 | |
| D90242.1 | - | 3 - 4 | 635 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkch
Genomic Map