Assay Details
| Gene Symbol: Slamf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013730.4 | NP_038758.2 | 5 - 6 | 982 | 128 | |
| GenBank mRNA | AB196796.1 | - | 5 - 6 | 887 | 128 | |
| AB196797.1 | - | 5 - 6 | 887 | 128 | ||
| AB196798.1 | - | 5 - 6 | 887 | 128 | ||
| AB196799.1 | - | 5 - 6 | 887 | 128 | ||
| AF149791.1 | - | 5 - 6 | 947 | 128 | ||
| AF160990.1 | - | 5 - 6 | 953 | 128 | ||
| AK016818.1 | - | 4 - 5 | 656 | 128 | ||
| AK020012.1 | - | 5 - 6 | 983 | 128 | ||
| AK036429.1 | - | 5 - 6 | 958 | 128 | ||
| AK156445.1 | - | 5 - 6 | 984 | 128 | ||
| AK169547.1 | - | 3 - 4 | 263 | 128 | ||
| BC117095.1 | - | 5 - 6 | 923 | 40126414 | 128 | |
| BC117099.1 | - | 5 - 6 | 923 | 40126418 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slamf1
Genomic Map