Assay Details
| Gene Symbol: Ifi203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK137616.1 | - | 5 - 6 | 678 | 59 | |
| AK144663.1 | - | 5 - 6 | 660 | 59 | ||
| BC008167.1 | - | 781 | 3257492 | 59 | ||
| CA574467.1 | - | 3 - 4 | 395 | 59 | ||
| L14559.1 | - | 1 - 2 | 171 | 59 | ||
More Information
Set Membership: |
|

Ifi203
Genomic Map