Assay Details
| Gene Symbol: Pex19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023041.3 | NP_075528.3 | 1 - 2 | 100 | 59 | |
| GenBank mRNA | AK029368.1 | - | 1 - 2 | 101 | 59 | |
| AK075576.1 | - | 1 - 2 | 87 | 59 | ||
| AK170175.1 | - | 1 - 2 | 89 | 59 | ||
| BC019767.1 | - | 1 - 2 | 83 | 4502819 | 59 | |
| CT010305.1 | - | 1 - 2 | 76 | 59 | ||
| Y09046.1 | - | 1 - 2 | 81 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pex19
Genomic Map