Assay Details
| Gene Symbol: Pea15a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011063.2 | NP_035193.1 | 1 - 2 | 190 | 60 | |
| GenBank mRNA | AK077421.1 | - | 1 - 2 | 119 | 60 | |
| AK089070.1 | - | 1 - 2 | 118 | 60 | ||
| AK153141.1 | - | 1 - 2 | 123 | 60 | ||
| AK153493.1 | - | 1 - 2 | 121 | 60 | ||
| AK161635.1 | - | 1 - 2 | 106 | 60 | ||
| AK161977.1 | - | 1 - 2 | 119 | 60 | ||
| BC038282.1 | - | 1 - 2 | 98 | 4500957 | 60 | |
| BY122161.1 | - | 1 - 2 | 191 | 60 | ||
| G49234.1 | - | 1 - 2 | 112 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK089070.1, AK153141.1, AK153493.1, AK161635.1, AK161977.1, BC038282.1, NM_011063.2 transcripts.
Set Membership: |
|

Pea15a
Genomic Map