Assay Details
| Gene Symbol: Vangl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033509.3 | NP_277044.1 | 4 - 5 | 1263 | 93 | |
| GenBank mRNA | AF365875.1 | - | 4 - 5 | 1027 | 93 | |
| AK046036.1 | - | 4 - 5 | 1280 | 93 | ||
| AK049613.1 | - | 1278 | 93 | |||
| AK129311.1 | - | 1263 | 93 | |||
| AY035370.3 | - | 4 - 5 | 1251 | 93 | ||
| BC052195.1 | - | 4 - 5 | 1263 | 6509008 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vangl2
Genomic Map