Assay Details
| Gene Symbol: Vangl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK129311.1 | - | 1403 | 60 | ||
| BC052195.1 | - | 5 - 6 | 1403 | 6509008 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vangl2
Genomic Map