Assay Details
| Gene Symbol: Kdsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027534.2 | NP_081810.1 | 3 - 4 | 348 | 108 | |
| GenBank mRNA | AK018155.1 | - | 4 - 5 | 323 | 108 | |
| AY634684.1 | - | 3 - 4 | 257 | 108 | ||
| BC023820.1 | - | 3 - 4 | 338 | 5354804 | 108 | |
More Information
Set Membership: |
|

Kdsr
Genomic Map