Assay Details
| Gene Symbol: Kdsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027534.2 | NP_081810.1 | 8 - 9 | 875 | 66 | |
| GenBank mRNA | AK018155.1 | - | 9 - 10 | 850 | 66 | |
| AK170064.1 | - | 8 - 9 | 758 | 66 | ||
| AY634684.1 | - | 8 - 9 | 784 | 66 | ||
| BC023820.1 | - | 8 - 9 | 865 | 5354804 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kdsr
Genomic Map