Assay Details
| Gene Symbol: Ube2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC016117.1 | - | 2 - 3 | 359 | 4486610 | 56 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC016117.1 transcripts.
Set Membership: |
|

Ube2f
Genomic Map