Assay Details
| Gene Symbol: Agxt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276710.1 | NP_001263639.1 | 7 - 8 | 891 | 81 | |
| NM_016702.3 | NP_057911.2 | 7 - 8 | 891 | 81 | ||
| GenBank mRNA | AF027730.2 | - | 7 - 8 | 886 | 81 | |
| AK149432.1 | - | 7 - 8 | 814 | 81 | ||
| BC025799.1 | - | 7 - 8 | 855 | 5099627 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Agxt
Genomic Map