Assay Details
| Gene Symbol: Gpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016696.4 | NP_057905.1 | 7 - 8 | 1510 | 60 | |
| GenBank mRNA | AF185613.1 | - | 7 - 8 | 1513 | 60 | |
| AK154584.1 | - | 5 - 6 | 644 | 60 | ||
| AK154899.1 | - | 7 - 8 | 1553 | 60 | ||
| AK206330.1 | - | 2 - 3 | 201 | 60 | ||
| BC039232.1 | - | 3 - 4 | 317 | 60 | ||
| BC062902.1 | - | 7 - 8 | 1339 | 6810413 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpc1
Genomic Map