Assay Details
| Gene Symbol: Armc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027456.1 | NP_081732.1 | 16 - 17 | 1626 | 61 | |
| NM_030184.2 | NP_084460.1 | 16 - 17 | 1661 | 61 | ||
| GenBank mRNA | AK017564.1 | - | 16 - 17 | 1627 | 61 | |
| AK019600.1 | - | 16 - 17 | 1650 | 61 | ||
| AK030307.1 | - | 16 - 17 | 1658 | 61 | ||
| AK036083.1 | - | 16 - 17 | 1632 | 61 | ||
| AK039800.1 | - | 16 - 17 | 1634 | 61 | ||
| AK045699.1 | - | 16 - 17 | 1625 | 61 | ||
| AK046693.1 | - | 16 - 17 | 1644 | 61 | ||
| AK140598.1 | - | 16 - 17 | 1657 | 61 | ||
| AK161393.1 | - | 16 - 17 | 1641 | 61 | ||
| AK164041.1 | - | 16 - 17 | 1660 | 61 | ||
| AK181387.1 | - | 2 - 3 | 61 | |||
| BC028964.1 | - | 9 - 10 | 881 | 5364429 | 61 | |
| BC043101.1 | - | 16 - 17 | 1613 | 6406321 | 61 | |
| BQ173978.1 | - | 5 - 6 | 61 | |||
More Information
Set Membership: |
|

Armc9
Genomic Map