Assay Details
| Gene Symbol: Fmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018881.3 | NP_061369.2 | 1 - 2 | 38 | 68 | |
| GenBank mRNA | AK009224.1 | - | 1 - 2 | 73 | 68 | |
| BC031415.1 | - | 1 - 2 | 38 | 3990305 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC031415.1, NM_018881.3 transcripts.
Set Membership: |
|

Fmo2
Genomic Map