Assay Details
| Gene Symbol: Fmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018881.3 | NP_061369.2 | 5 - 6 | 677 | 103 | |
| GenBank mRNA | AF184981.1 | - | 4 - 5 | 742 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fmo2
Genomic Map