Assay Details
| Gene Symbol: Fmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018881.3 | NP_061369.2 | 6 - 7 | 867 | 93 | |
| GenBank mRNA | AF184981.1 | - | 5 - 6 | 932 | 93 | |
| AK129026.1 | - | 2 - 3 | 400 | 93 | ||
| BC031415.1 | - | 6 - 7 | 867 | 3990305 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fmo2
Genomic Map