Assay Details
| Gene Symbol: Sft2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145512.4 | NP_663487.1 | 4 - 5 | 448 | 60 | |
| GenBank mRNA | AK036578.1 | - | 4 - 5 | 336 | 60 | |
| AK075812.1 | - | 3 - 4 | 278 | 60 | ||
| AK156973.1 | - | 4 - 5 | 339 | 60 | ||
| AK170590.1 | - | 3 - 4 | 253 | 60 | ||
| BC017549.1 | - | 4 - 5 | 327 | 4924681 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sft2d2
Genomic Map