Assay Details
| Gene Symbol: Mpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008623.4 | NP_032649.2 | 6 - 7 | 751 | 114 | |
| GenBank mRNA | AK089180.1 | - | 6 - 7 | 755 | 114 | |
| AK157960.1 | - | 4 - 5 | 648 | 114 | ||
| BC139139.1 | - | 4 - 5 | 616 | 8862161 | 114 | |
| BC141226.1 | - | 4 - 5 | 616 | 9055838 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mpz
Genomic Map