Assay Details
| Gene Symbol: Blzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160208.1 | NP_001153680.1 | 5 - 6 | 964 | 80 | |
| NM_001160209.1 | NP_001153681.1 | 4 - 5 | 1051 | 80 | ||
| NM_025505.4 | NP_079781.2 | 6 - 7 | 985 | 80 | ||
| GenBank mRNA | AK005489.1 | - | 4 - 5 | 785 | 80 | |
| AK006544.1 | - | 5 - 6 | 968 | 80 | ||
| AK029787.1 | - | 6 - 7 | 986 | 80 | ||
| AK167493.1 | - | 4 - 5 | 1058 | 80 | ||
| AK168933.1 | - | 4 - 5 | 812 | 80 | ||
| AK171167.1 | - | 4 - 5 | 843 | 80 | ||
| BC027025.1 | - | 4 - 5 | 826 | 5374749 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Blzf1
Genomic Map