Assay Details
| Gene Symbol: Tomm40l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037170.2 | NP_001032247.1 | 5 - 6 | 660 | 62 | |
| GenBank mRNA | AK012264.1 | - | 5 - 6 | 590 | 62 | |
| AK028388.1 | - | 5 - 6 | 661 | 62 | ||
| AK029485.1 | - | 2 - 3 | 156 | 62 | ||
| AK186544.1 | - | 3 - 4 | 238 | 62 | ||
| AK190108.1 | - | 2 - 3 | 146 | 62 | ||
| AK207572.1 | - | 2 - 3 | 146 | 62 | ||
| BC058978.1 | - | 4 - 5 | 551 | 5715769 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2032363 | BC058978.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2032363 | AK140721.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2032363 | AK028388.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2032363 | NM_001037170.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2032363 | AK149381.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 572276 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 64410 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 172002 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179523 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2032363 | BC058978.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2032363 | AK140721.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2032363 | AK028388.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2032363 | NM_001037170.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2032363 | AK149381.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826709 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826724 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826702 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826734 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826720 | - | - |
| Whole Mouse Genome G4122A | A_52_P86003 | BC058978.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P86003 | AK177299.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P86003 | AK178629.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P86003 | AK208296.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P86003 | AK028388.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134523 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00914434 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01015401 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00868373 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00998717 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01055836 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00868357 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00868402 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00998722 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00868342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826739 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00868374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00868399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00868357 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826724 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00868402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01055833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826693 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826725 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00868353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826729 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00868342 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826710 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826727 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00868412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826686 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10360059 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360066 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360069 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360062 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351545 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360068 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360064 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360061 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360060 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360063 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360067 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360065 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455614_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429382_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351533 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10360058 | - | - |
| Mouse Genome 430 2.0 Array | 1455614_at | - | - |
| Mouse Genome 430 2.0 Array | 1429382_at | - | - |
| Mouse Genome 430 2.0 Array | 1455614_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429382_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 641376_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 641376_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100028884.mm.1 | NM_001037170.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028884.mm.1 | NM_001037170.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028884.mm.1 | TC0100003532.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100028885.mm.1 | NM_001037170.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028885.mm.1 | NM_001037170.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100028885.mm.1 | TC0100003532.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429382_at | - | - |
| Mouse Expression Set 430 | 1429382_PM_at | - | - |
| Mouse Expression Set 430 | 1455614_PM_at | - | - |
| Mouse Expression Set 430 | 1455614_at | - | - |
| Murine Genome U74v2 Set | 168127_at | - | - |
| Murine Genome U74v2 Set | 168127_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429382_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429382_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429382_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429382_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455614_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455614_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 723965 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 105080114 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105080114 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106840154 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2588846 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1212992 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2588846 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1212992 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2481771 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221049 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232024 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254075 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106840154 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105080114 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105080114 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106840154 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254075 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1232024 | - | - |

Tomm40l
Genomic Map