Assay Details
| Gene Symbol: Arhgap30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005508.2 | NP_001005508.2 | 10 - 11 | 1704 | 89 | |
| GenBank mRNA | AK031319.1 | - | 10 - 11 | 1475 | 89 | |
| AK147757.1 | - | 10 - 11 | 1712 | 89 | ||
| AK156351.1 | - | 11 | 1718 | 113 | ||
| AK220204.1 | - | 6 | 862 | 113 | ||
| BC082573.1 | - | 10 - 11 | 1652 | 30548035 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap30
Genomic Map