Assay Details
| Gene Symbol: Nckap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081756.1 | NP_001075225.1 | 2 - 3 | 445 | 86 | |
| NM_176957.3 | NP_795931.2 | 2 - 3 | 445 | 86 | ||
| GenBank mRNA | AK041950.1 | - | 2 - 3 | 295 | 86 | |
| AK083792.1 | - | 2 - 3 | 383 | 86 | ||
| CN700744.1 | - | 2 - 3 | 455 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041950.1 transcripts.
Set Membership: |
|

Nckap5
Genomic Map