Assay Details
| Gene Symbol: Nckap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081756.1 | NP_001075225.1 | 3 - 4 | 516 | 80 | |
| NM_176957.3 | NP_795931.2 | 3 - 4 | 516 | 80 | ||
| GenBank mRNA | CN700744.1 | - | 3 - 4 | 526 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nckap5
Genomic Map