Assay Details
| Gene Symbol: Nckap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081756.1 | NP_001075225.1 | 7 - 8 | 957 | 58 | |
| NM_172484.3 | NP_766072.2 | 5 - 6 | 818 | 58 | ||
| NM_176957.3 | NP_795931.2 | 7 - 8 | 957 | 58 | ||
| GenBank mRNA | AK053243.1 | - | 5 - 6 | 640 | 58 | |
| AY957601.1 | - | 3 - 4 | 377 | 58 | ||
| AY957602.1 | - | 3 - 4 | 377 | 58 | ||
| BC066095.1 | - | 5 - 6 | 818 | 6856184 | 58 | |
| BC079551.1 | - | 5 - 6 | 818 | 6856184 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nckap5
Genomic Map